L(+)-Lysin Dihydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-Lys-OH.2HCl, (S)-(+)-2,6-Diaminohexansäure Dihydrochlorid, (S)-(+)-2,6-Diaminocapronsäure Dihydrochlorid, (S)-(+)-Lysin Dihydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

L5751-100G
SIALL5751-100GEA 206 EUR
SIALL5751-100G
L(+)-Lysin Dihydrochlorid, Sigma-Aldrich®
L(+)-Lysin Dihydrochlorid
Formel: C₆H₁₆Cl₂N₂O₂
Molecular Weight: 219.11 g/mol
Schmelzpunkt: 200,2
MDL: MFCD00066389
CAS-Nummer: 657-26-1
EINECS: 211-518-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR