DL-Lysin Monohydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(±)-Lysin Hydrochlorid, (±)-Lysin Monohydrochlorid, (±)-2,6-Diaminohexansäure Hydrochlorid, (±)-2,6-Diaminocapronsäure Monohydrochlorid, (±)-2,6-Diaminohexansäure Monohydrochlorid, DL-Lysin Hydrochlorid, H-DL-Lys-OH.HCl, (±)-2,6-Diaminocapronsäure Hydrochlorid
M-Clarity™ Qualitätsstufe = MQ200
L6001-100G
L6001-1KG
L6001-25G
SIALL6001-100GEA
146
EUR
SIALL6001-100G
SIALL6001-1KG
SIALL6001-25G
DL-Lysin Monohydrochlorid, Sigma-Aldrich®
DL-Lysin Monohydrochlorid
Formel:
C₆H₁₅ClN₂O₂ Schmelzpunkt: 267 °C |
MDL:
MFCD00064563 CAS-Nummer: 70-53-1 EINECS: 200-739-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...