DL-Lysin Monohydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (±)-Lysin Hydrochlorid, (±)-Lysin Monohydrochlorid, (±)-2,6-Diaminohexansäure Hydrochlorid, (±)-2,6-Diaminocapronsäure Monohydrochlorid, (±)-2,6-Diaminohexansäure Monohydrochlorid, DL-Lysin Hydrochlorid, H-DL-Lys-OH.HCl, (±)-2,6-Diaminocapronsäure Hydrochlorid

M-Clarity™ Qualitätsstufe = MQ200

L6001-100G L6001-1KG L6001-25G
SIALL6001-100GEA 146 EUR
SIALL6001-100G SIALL6001-1KG SIALL6001-25G
DL-Lysin Monohydrochlorid, Sigma-Aldrich®
DL-Lysin Monohydrochlorid
Formel: C₆H₁₅ClN₂O₂
Schmelzpunkt: 267 °C
MDL: MFCD00064563
CAS-Nummer: 70-53-1
EINECS: 200-739-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR