D(-)-Luciferin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: D-Luciferin, (S)-2-(6-Hydroxybenzo[d]thiazol-2-yl)-4,5-dihydrothiazole-4-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ200

L6152-10MG L6152-1MG
SIALL6152-10MGEA 1130 EUR
SIALL6152-10MG SIALL6152-1MG
D(-)-Luciferin, Sigma-Aldrich®
D(-)-Luciferin
Formel: C₁₁H₈N₂O₃S₂
MDL: MFCD00042929
CAS-Nummer: 2591-17-5
EINECS: 219-981-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR