3-Methoxybenzamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: m-Anisamid, 3-Methoxybenzoesäureamid

M-Clarity™ Qualitätsstufe = MQ100

M10050-5G
SIALM10050-5GEA 85.8 EUR
SIALM10050-5G
3-Methoxybenzamid, Sigma-Aldrich®
3-Methoxybenzamid
Formel: C₈H₉NO₂
Molecular Weight: 151,16 g/mol
MDL: MFCD00007986
CAS-Nummer: 5813-86-5
EINECS: 227-379-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR