5-Methoxygramin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-(5-Methoxy-1H-indol-3-yl)-N,N-dimethylmethanamine

M-Clarity™ Qualitätsstufe = MQ100

M14870-5G
SIALM14870-5GEA 225 EUR
SIALM14870-5G
5-Methoxygramin, Sigma-Aldrich®
5-Methoxygramin
Formel: C₁₂H₁₆N₂O
Molecular Weight: 204,27 g/mol
MDL: MFCD00005630
CAS-Nummer: 16620-52-3
EINECS: 240-669-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR