5-Methoxy-3-indolylessigsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 5-Methoxyindol-3-essigsäure, 5-Methoxyindol-3-ylessigsäure

M-Clarity™ Qualitätsstufe = MQ100

M14935-1G
SIALM14935-1GEA 143 EUR
SIALM14935-1G
5-Methoxy-3-indolylessigsäure, Sigma-Aldrich®
5-Methoxy-3-indolylessigsäure
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,21 g/mol
Schmelzpunkt: 150…151 °C
MDL: MFCD00005638
CAS-Nummer: 3471-31-6
EINECS: 222-438-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR