4-Methyl-m-anissäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Methoxy-4-methylbenzoesäure

M-Clarity™ Qualitätsstufe = MQ100

M15052-5G
SIALM15052-5GEA 125 EUR
SIALM15052-5G
4-Methyl-m-anissäure, Sigma-Aldrich®
4-Methyl-m-anissäure
Formel: C₉H₁₀O₃
Molecular Weight: 166,18 g/mol
MDL: MFCD00002566
CAS-Nummer: 7151-68-0
EINECS: 230-486-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR