1-(4-Methoxyphenyl)piperazin Dihydrochlorid, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
M23004-25G
SIALM23004-25GEA
96.7
EUR
SIALM23004-25G
1-(4-Methoxyphenyl)piperazin Dihydrochlorid, Sigma-Aldrich®
1-(4-Methoxyphenyl)piperazin Dihydrochlorid
Formel:
C₁₁H₁₈Cl₂N₂O Molecular Weight: 265,18 g/mol |
MDL:
MFCD00012767 CAS-Nummer: 38869-47-5 EINECS: 254-166-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...