3'-Methylacetophenon, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Methyl-m-tolylketon, m-Methylacetophenon

M-Clarity™ Qualitätsstufe = MQ200

M26607-25G M26607-5G
SIALM26607-25GEA 96 EUR
SIALM26607-25G SIALM26607-5G
3'-Methylacetophenon, Sigma-Aldrich®
3'-Methylacetophenon
Formel: C₉H₁₀O
Molecular Weight: 134.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00008742
CAS-Nummer: 585-74-0
EINECS: 209-561-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR