(Methylamino)acetaldehyddimethylacetal, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2,2-Dimethoxy-N-methylethanamine

MQ100

M28006-25G
SIALM28006-25GEA 48.5 EUR
SIALM28006-25G
(Methylamino)acetaldehyddimethylacetal, Sigma-Aldrich®
(Methylamino)acetaldehyddimethylacetal
Formel: C₅H₁₃NO₂
Molecular Weight: 119.16 g/mol
MDL: MFCD00008485
CAS-Nummer: 122-07-6
EINECS: 204-520-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR