Methyl-4-chlorbutyrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl 4-chlorobutyrate

M-Clarity™ Qualitätsstufe = MQ100

M34804-100G
SIALM34804-100GEA 58.3 EUR
SIALM34804-100G
Methyl-4-chlorbutyrat, Sigma-Aldrich®
Methyl-4-chlorbutyrat
Formel: C₅H₉ClO₂
Molecular Weight: 136.58 g/mol
MDL: MFCD00001003
CAS-Nummer: 3153-37-5
EINECS: 221-592-9
UN: 3272
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR