2-Mercaptobenzoxazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Benzo[d]oxazole-2-thiol

M-Clarity™ Qualitätsstufe = MQ200

M3507-10G
SIALM3507-10GEA 31.5 EUR
SIALM3507-10G
2-Mercaptobenzoxazol, Sigma-Aldrich®
2-Mercaptobenzoxazole
Formel: C₇H₅NOS
Molecular Weight: 151,19 g/mol
MDL: MFCD00005769
CAS-Nummer: 2382-96-9
EINECS: 219-191-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR