(R)-(+)-3-Methylcyclopentanon, Sigma-Aldrich®

Lieferant: Merck

M39709-1G
SIALM39709-1GEA 112 EUR
SIALM39709-1G
(R)-(+)-3-Methylcyclopentanon, Sigma-Aldrich®
(R)-(+)-3-Methylcyclopentanon
Formel: C₆H₁₀O
Molecular Weight: 98,14 g/mol
CAS-Nummer: 6672-30-6
EINECS: 229-711-6
UN: 1224
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR