3-Methylglutarsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

M47604-25G
SIALM47604-25GEA 132 EUR
SIALM47604-25G
3-Methylglutarsäure, Sigma-Aldrich®
3-Methylglutarsäure
Formel: C₆H₁₀O₄
Molecular Weight: 146,1412 g/mol
MDL: MFCD00002727
CAS-Nummer: 626-51-7
EINECS: 210-951-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR