Dinatriummalonat Monohydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium malonate dibasic monohydrate, Malonic acid disodium salt hydrate

M-Clarity™ Qualitätsstufe = MQ100

M4795-25G
SIALM4795-25GEA 112 EUR
SIALM4795-25G
Dinatriummalonat Monohydrat, Sigma-Aldrich®
Dinatriummalonat Monohydrat
Formel: C₃H₂Na₂O₄·1H₂O
Molecular Weight: 166.04 g/mol
MDL: MFCD00149114
CAS-Nummer: 26522-85-0
EINECS: 205-514-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR