Methylisonicotinat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Pyridinecarboxylic acid methyl ester, 4-Picolinic acid methyl ester, Methyl 4-pyridinecarboxylate, Pyridine-4-carboxylic acid methyl ester, Isonicotinic acid methyl ester

M-Clarity™ Qualitätsstufe = MQ100

M52950-100G
SIALM52950-100GEA 73.4 EUR
SIALM52950-100G
Methylisonicotinat, Sigma-Aldrich®
Methylisonicotinat
Formel: C₇H₇NO₂
Molecular Weight: 137,14 g/mol
Siedepunkt: 207 °C (1013 hPa)
Schmelzpunkt: 17 °C
Dichte: 1,161 g/cm³ (20 °C)
MDL: MFCD00006427
CAS-Nummer: 2459-09-8
EINECS: 219-546-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR