5-Methoxytryptamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-(5-Methoxy-1H-indol-3-yl)ethanamine, O-Methylserotonin hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

M6628-1G
SIALM6628-1GEA 295 EUR
SIALM6628-1G
5-Methoxytryptamin Hydrochlorid, Sigma-Aldrich®
5-Methoxytryptamin Hydrochlorid
Formel: C₁₁H₁₅ClN₂O
MDL: MFCD00005662
CAS-Nummer: 66-83-1
EINECS: 200-637-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR