(±)-1-Methyl-3-phenylpropylamin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3-Amino-1-phenylbutane, 3-Amino-1-phenyl-butane

M-Clarity™ Qualitätsstufe = MQ100

M70533-25G
SIALM70533-25GEA 91.7 EUR
SIALM70533-25G
(±)-1-Methyl-3-phenylpropylamin, Sigma-Aldrich®
(±)-1-Methyl-3-phenylpropylamin
Formel: C₁₀H₁₅N
Molecular Weight: 149,24 g/mol
MDL: MFCD00008090
CAS-Nummer: 22374-89-6
EINECS: 244-942-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR