2-Formyl-6-picolin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

M78208-25G M78208-5G
SIALM78208-25GEA 1030 EUR
SIALM78208-25G SIALM78208-5G
2-Formyl-6-picolin, Sigma-Aldrich®
2-Formyl-6-picolin
Formel: C₇H₇NO
MDL: MFCD00006291
CAS-Nummer: 1122-72-1
EINECS: 214-359-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR