1-Methyl-2-tetralon, Sigma-Aldrich®

Lieferant: Merck

M82906-5G
SIALM82906-5GEA 198 EUR
SIALM82906-5G
1-Methyl-2-tetralon, Sigma-Aldrich®
1-Methyl-2-tetralon
Formel: C₁₁H₁₂O
Molecular Weight: 160,22 g/mol
CAS-Nummer: 4024-14-0
EINECS: 223-690-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR