2-Methyl-2-thiazolin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Methyl-4,5-dihydrothiazole

M-Clarity™ Qualitätsstufe = MQ100

M83406-25G
SIALM83406-25GEA 70.6 EUR
SIALM83406-25G
2-Methyl-2-thiazolin, Sigma-Aldrich®
2-Methyl-2-thiazolin
Formel: C₄H₇NS
Molecular Weight: 101,17 g/mol
MDL: MFCD00005314
CAS-Nummer: 2346-00-1
EINECS: 219-071-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR