5-Methylthiophen-2-carbaldehyd, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

M84410-25G
SIALM84410-25GEA 41.3 EUR
SIALM84410-25G
5-Methylthiophen-2-carbaldehyd, Sigma-Aldrich®
5-Methylthiophen-2-carbaldehyd
Formel: C₆H₆OS
MDL: MFCD00005434
CAS-Nummer: 13679-70-4
EINECS: 237-178-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR