Methyl-3,4,5-trimethoxybenzoat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 3,4,5-Trimethoxybenzoesäuremethylester

M-Clarity™ Qualitätsstufe = MQ100

M86006-100G
SIALM86006-100GEA 50.2 EUR
SIALM86006-100G
Methyl-3,4,5-trimethoxybenzoat, Sigma-Aldrich®
Methyl-3,4,5-trimethoxybenzoat
Formel: C₁₁H₁₄O₅
Molecular Weight: 226,23 g/mol
MDL: MFCD00008431
CAS-Nummer: 1916-07-0
EINECS: 217-629-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR