Dinatriummaleat Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Sodium toxilate hydrate, Sodium maleate xhydrate, Maleic acid disodium salt hydrate

M-Clarity™ Qualitätsstufe = MQ200

M9009-100G
SIALM9009-100GEA 176 EUR
SIALM9009-100G
Dinatriummaleat Hydrat, Sigma-Aldrich®
Dinatriummaleat Hydrat
Formel: C₄H₂Na₂O₄·xH₂O
Molecular Weight: 160.04 g/mol (anhy)
MDL: MFCD00150133
CAS-Nummer: 25880-69-7
EINECS: 206-738-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR