Methoxyacetonitril, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
M9009-100G-A
M9009-25G-A
M9009-5G-A
SIALM9009-100G-AEA
588
EUR
SIALM9009-100G-A
SIALM9009-25G-A
SIALM9009-5G-A
Methoxyacetonitril, Sigma-Aldrich®
Methoxyacetonitril
Formel:
C₃H₅NO |
MDL:
MFCD00001892 CAS-Nummer: 1738-36-9 EINECS: 217-092-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...