High Temperature Viscosity Standards, Supelco®
Lieferant: Merck
N100HT-500ML
SIALN100HT-500MLEA
222
EUR
SIALN100HT-500ML
High Temperature Viscosity Standards, Supelco®
Standards
Viskosimetrie-Standards
- Temperature range: 100 to 150 °C
- Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/ml) given at all temperatures
- Fully traceable to National Standards
- Ensures full compliance to ASTM & IP test method protocol
- Tamper evident security packaging
Standards: Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation.