(2-Nitrophenyl)-β-D-galactopyranosid (ONPG), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
o-Nitrophenyl-β-D-galactoside, 2-Nitrophenyl β-D-Galactopyranoside, O-Nitrophenyl-Β-D-Galactopyranoside, ONPG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(2-nitrophenoxy)tetrahydro-2H-pyran-3,4,5-triol
M-Clarity™ Qualitätsstufe = MQ200
N1127-1G
N1127-250MG
N1127-25G
N1127-500MG
N1127-5G
SIALN1127-1GEA
56.4
EUR
SIALN1127-1G
SIALN1127-250MG
SIALN1127-25G
SIALN1127-500MG
SIALN1127-5G
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG), Sigma-Aldrich®
(2-Nitrophenyl)-β-D-galactopyranosid (ONPG)
Formel:
C₁₂H₁₅NO₈ Lagertemperatur: Gefrierschrank |
MDL:
MFCD00063255 CAS-Nummer: 369-07-3 EINECS: 206-716-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...