β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂) Hydrat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: beta-NADH disodium salt hydrate, beta-Nicotinamide adenine dinucleotide disodium salt hydrate, β-Dihydronicotinamide adenine dinucleotide disodium salt (NADH-Na2, reduced form) hydrate

N1161-10VL
SIALN1161-10VLEA 423 EUR
SIALN1161-10VL
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂) Hydrat, Sigma-Aldrich®
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na2) Hydrat
Formel: C₂₁H₂₇N₇Na₂O₁₄P₂·H₂O
Molecular Weight: 727,43 g/mol
MDL: MFCD03791273
CAS-Nummer: 606-68-8
EINECS: 210-123-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR