ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz (reduzierte Form)
M-Clarity™ Qualitätsstufe = MQ300
N1630-25MG
SIALN1630-25MGEA
154
EUR
SIALN1630-25MG
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4), Sigma-Aldrich®
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4)
Formel:
C₂₁H₂₆N₇Na₄O₁₇P₃ Molecular Weight: 833,35 g/mol |
MDL:
MFCD00036263 CAS-Nummer: 2646-71-1 EINECS: 220-163-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...