6-Nitro-1H-indazol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-NINA, 6-Nitro-1H-indazol

M-Clarity™ Qualitätsstufe = MQ100

N17513-100G
SIALN17513-100GEA 220 EUR
SIALN17513-100G
6-Nitro-1H-indazol, Sigma-Aldrich®
6-Nitro-1H-indazole
Formel: C₇H₅N₃O₂
Molecular Weight: 163,14 g/mol
Schmelzpunkt: 181 °C
MDL: MFCD00005695
CAS-Nummer: 7597-18-4
EINECS: 231-500-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR