Bis(4-nitrophenyl)disulfid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,2-Bis(4-nitrophenyl)disulphane, 4-Nitrophenyl disulphide, 4,4'-Dinitrodiphenyl Disulphide

M-Clarity™ Qualitätsstufe = MQ100

N21022-25G
SIALN21022-25GEA 72.9 EUR
SIALN21022-25G
Bis(4-nitrophenyl)disulfid, Sigma-Aldrich®
Bis(4-nitrophenyl)disulfid
Formel: C₁₂H₈N₂O₄S₂
Molecular Weight: 308,34 g/mol
MDL: MFCD00003573
CAS-Nummer: 100-32-3
EINECS: 202-840-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR