Potassium-4-nitrophenyl sulphate ≥98% (durch TLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Nitrophenyl sulphate, potassium salt, Potassium 4-Nitrophenyl sulphate, 4-Nitrophenyl sulphate potassium salt, sulphuric acid 4-Nitrophenyl ester potassium salt

M-Clarity™ Qualitätsstufe = MQ200

N3877-1G N3877-500MG N3877-5G
SIALN3877-1GEA 206 EUR
SIALN3877-1G SIALN3877-500MG SIALN3877-5G
Potassium-4-nitrophenyl sulphate ≥98% (durch TLC), Sigma-Aldrich®
Potassium-4-nitrophenyl sulphate

Chromogenic sulphatase substrate.


Solubility in water 50 mg/ml, clear.

Formel: C₆H₄KNO₆S
Schmelzpunkt: 246…250 °C (Lit.)
Lagertemperatur: Gefrierschrank
MDL: MFCD00007466
CAS-Nummer: 6217-68-1
EINECS: 228-288-5

Bestellen

Spezifikation Testergebnisse

Carbon 27.4 - 28.6%
Nitrogen 27.4 - 28.6%
Free p-Nitrophenol ≤0.02%
Potassium (K) 14 - 16%
Purity (TLC) ≥98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR