Nystatin, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Fungicidin, Mycostatin

M-Clarity™ Qualitätsstufe = MQ300

N4014-50MG
SIALN4014-50MGEA 92.6 EUR
SIALN4014-50MG
Nystatin, Sigma-Aldrich®
Nystatin
Formel: C₄₇H₇₅NO₁₇
Molecular Weight: 926,09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD25541651
CAS-Nummer: 1400-61-9
EINECS: 215-749-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR