Nonadecansäuremethylester, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Nonadecanoic acid methyll ester, Methyl Nonadecanoate

M-Clarity™ Qualitätsstufe = MQ200

N5377-1G N5377-10G N5377-5G
SIALN5377-1GEA 108 EUR
SIALN5377-1G SIALN5377-10G SIALN5377-5G
Nonadecansäuremethylester, Sigma-Aldrich®
Nonadecanoic acid methyl ester
Formel: C₂₀H₄₀O₂
Molecular Weight: 312.54 g/mol
MDL: MFCD00009011
CAS-Nummer: 1731-94-8
EINECS: 217-056-9

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR