2-(1-Naphthylmethyl)-2-imidazolin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Naphazoline Hydrochloride, Albalon

M-Clarity™ Qualitätsstufe = MQ100

N5504-25G
SIALN5504-25GEA 69.1 EUR
SIALN5504-25G
2-(1-Naphthylmethyl)-2-imidazolin Hydrochlorid, Sigma-Aldrich®
2-(1-Naphthylmethyl)-2-imidazolin Hydrochlorid
Formel: C₁₄H₁₅ClN₂
Molecular Weight: 246,738 g/mol
MDL: MFCD00012554
CAS-Nummer: 550-99-2
EINECS: 208-989-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR