(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on, Sigma-Aldrich®

Lieferant: Merck

N5893-1G N5893-25G N5893-5G
SIALN5893-1GEA 53.3 EUR
SIALN5893-1G SIALN5893-25G SIALN5893-5G
(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on, Sigma-Aldrich®
(±)-Naringenin
Formel: C₁₅H₁₂O₅
MDL: MFCD00006844
CAS-Nummer: 67604-48-2
EINECS: 266-769-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR