(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on, Sigma-Aldrich®
Lieferant: Merck
N5893-1G
N5893-25G
N5893-5G
SIALN5893-1GEA
53.3
EUR
SIALN5893-1G
SIALN5893-25G
SIALN5893-5G
(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on, Sigma-Aldrich®
(±)-Naringenin
Formel:
C₁₅H₁₂O₅ |
MDL:
MFCD00006844 CAS-Nummer: 67604-48-2 EINECS: 266-769-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...