β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂), Sigma-Aldrich®
Lieferant: Merck
N6660-15VL
SIALN6660-15VLEA
258
EUR
SIALN6660-15VL
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na₂), Sigma-Aldrich®
β-Nicotinamid-Adenin-Dinucleotid Dinatriumsalz, reduzierte Form (NADH-Na2)
Formel:
C₂₁H₂₇N₇Na₂O₁₄P₂ |
MDL:
MFCD00036200 CAS-Nummer: 606-68-8 EINECS: 210-123-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...