DL-Norvalin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
(±)-2-Aminovaleriansäure, (±)-Norvalin, H-DL-Nva-OH, (±)-2-Aminopentansäure
M-Clarity™ Qualitätsstufe = MQ200
N7502-25G
SIALN7502-25GEA
64.3
EUR
SIALN7502-25G
DL-Norvalin, Sigma-Aldrich®
DL-Norvalin
Formel:
C₅H₁₁NO₂ Molecular Weight: 117,15 g/mol Siedepunkt: 223 °C (1013 hPa) Schmelzpunkt: > 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064420 CAS-Nummer: 760-78-1 EINECS: 212-082-7 Merck Index: 12,06813 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...