4-Nitrophenyl phosphate disodium salt (pNNP) Substrat System, Flüssigkeit, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phosphoric acid 4-nitrophenyl ester disodium salt, 4-Nitrophenyl phosphate disodium salt, PNPP disodium, pNNP

M-Clarity™ Qualitätsstufe = MQ200

N7653-100ML
SIALN7653-100MLEA 252 EUR
SIALN7653-100ML
4-Nitrophenyl phosphate disodium salt (pNNP) Substrat System, Flüssigkeit, Sigma-Aldrich®
4-Nitrophenyl phosphate disodium salt (pNNP)

The p-Nitrophenyl Phosphate (pNPP) Liquid Substrate System combines p-nitrophenyl phosphate, buffer, and the required magnesium cations in a convenient, ready-to-use, single solution reagent. Producing a soluble end product, pNPP is the substrate of choice in alkaline phosphatase enzyme immunoassays.

Formel: C₆H₄NNa₂O₆P
Molecular Weight: 263,052 g/mol
Schmelzpunkt: 300 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00007319
CAS-Nummer: 4264-83-9
EINECS: 224-246-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR