ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na), Sigma-Aldrich®

Lieferant: Merck

Synonyme: NADP Mononatriumsalz, ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, NADP-Na, Nadidphosphat Mononatriumsalz, Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz

N8160-15VL
SIALN8160-15VLEA 538 EUR
SIALN8160-15VL
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form (NADP-Na), Sigma-Aldrich®
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Mononatriumsalz, oxidierte Form(NADP-Na)
Formel: C₂₁H₂₇N₇NaO₁₇P₃
Molecular Weight: 765.39 g/mol
MDL: MFCD00036973
CAS-Nummer: 1184-16-3
EINECS: 214-664-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR