ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz (reduzierte Form)

N9660-15VL
SIALN9660-15VLEA 517 EUR
SIALN9660-15VL
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4), Sigma-Aldrich®
ß-Nicotinamid-Adenin-Dinucleotid-Phosphat Tetranatriumsalz, reduzierte Form (NADPH-Na4)
Formel: C₂₁H₂₆N₇Na₄O₁₇P₃
Molecular Weight: 833,35 g/mol
MDL: MFCD00036263
CAS-Nummer: 2646-71-1
EINECS: 220-163-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR