di-Lithiumoxalat, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Dilithium oxalate, Lithium oxalate, Lithiumoxalate

O0130-500G
SIALO0130-500GEA 586 EUR
SIALO0130-500G
di-Lithiumoxalat, Sigma-Aldrich®
di-Lithiumoxalat
Formel: C₂Li₂O₄
Molecular Weight: 101,9 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00040596
CAS-Nummer: 553-91-3
EINECS: 209-054-1
UN: 3288
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR