L(+)-Ornithin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

O2375-100G O2375-10MG O2375-1KG O2375-25G O2375-5G
SIALO2375-100GEA 127 EUR
SIALO2375-100G SIALO2375-10MG SIALO2375-1KG SIALO2375-25G SIALO2375-5G
L(+)-Ornithin Hydrochlorid, Sigma-Aldrich®
L(+)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
MDL: MFCD00064562
CAS-Nummer: 3184-13-2
EINECS: 221-678-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR