D(-)-Ornithin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-D-Orn-OH·HCl, H-D-Orn-OH.HCl

M-Clarity™ Qualitätsstufe = MQ100

O5250-1G
SIALO5250-1GEA 61.1 EUR
SIALO5250-1G
D(-)-Ornithin Hydrochlorid, Sigma-Aldrich®
D(-)-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168.62 g/mol
MDL: MFCD00012917
CAS-Nummer: 16682-12-5
EINECS: 240-729-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR