N-Octyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-8, SB-8), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
3-(N,N-Dimethyloctylammonio)propanesulphonate inner salt
M-Clarity™ Qualitätsstufe = MQ200
O6626-5G
SIALO6626-5GEA
193
EUR
SIALO6626-5G
N-Octyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-8, SB-8), Sigma-Aldrich®
N-Octyl-N,N-dimethyl-3-ammonio-1-propansulfonat (Sulfobetain-8, SB-8)
Formel:
C₁₃H₂₉NO₃S Lagertemperatur: Raumtemperatur |
MDL:
MFCD00036912 CAS-Nummer: 15178-76-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...