Dioctylsulfid, Sigma-Aldrich®

Lieferant: Merck
Gefahr

Synonyme: Dioctylsulphane

M-Clarity™ Qualitätsstufe = MQ100

O7007-100ML-A
SIALO7007-100ML-AEA 131 EUR
SIALO7007-100ML-A
Dioctylsulfid, Sigma-Aldrich®
Dioctylsulfid
Formel: C₁₆H₃₄S
Molecular Weight: 258,51 g/mol
MDL: MFCD00009566
CAS-Nummer: 2690-08-6
EINECS: 220-259-5
UN: 3334
ADR: 9,

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR