17-Octadecinsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

O8382-25MG O8382-5MG
SIALO8382-25MGEA 1070 EUR
SIALO8382-25MG SIALO8382-5MG
17-Octadecinsäure, Sigma-Aldrich®
17-Octadecinsäure
Formel: C₁₈H₃₂O₂
Molecular Weight: 280.45 g/mol
CAS-Nummer: 34450-18-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR