7-Oxabicyclo[2.2.1]heptan, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

O8704-5G
SIALO8704-5GEA 106 EUR
SIALO8704-5G
7-Oxabicyclo[2.2.1]heptan, Sigma-Aldrich®
7-Oxabicyclo[2.2.1]heptan
Formel: C₆H₁₀O
Molecular Weight: 98,15 g/mol
MDL: MFCD00082342
CAS-Nummer: 279-49-2
EINECS: 205-998-3
UN: 3271
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR